Learn More
Medchemexpress LLC Ac-D-Ala-OH-d4 | 2708343-30-8 | 99.0% | 135.15 | 10 MG
SDP
Catalog No. 5000454857
Encompass_Preferred
Ac-D-Ala-OH-d4 is the deuterium labeled Ac-D-Ala-OH.
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
  • Product is for research use only
Catalog No. 50-004-54857 Supplier Medchemexpress LLC Supplier No. HYW012713S5MG
Ac-D-Ala-OH-d4 is the deuterium labeled Ac-D-Ala-OH.
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
  • Product is for research use only