Learn More
CPC Scientific Ac-Tyr-DArg-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific GHRF004A
Encompass
ONE-LETTER SEQUENCE: Ac-Y-DArg-DAIFTNSYRKVLGQLSARKLLQDIMSR-NH2MOLECULAR FORMULA: C154H255N47O43S1MOLECULAR WEIGHT:3485.08STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [93942-91-7], SYNONYMS: Acetyl-(D-Arg)-GRF (1-29) amide (human), Acetyl-(D-Arg)-SermorelinRESEARCH AREA: HormonalREFERENCES: P. Robberecht et al., The Endocrine Society, 117, 1759 (1985)
Catalog No. 50-210-9339