Learn More
Cayman Chemical Ac DArgCEH DPheRWCNH 10mg
SDP
Catalog No. 502943038
Encompass_Preferred
An MC4R agonist; induces cAMP release in HEK293 cells expressing human MC4R (EC50 = 0.28 nM); is selective for MC4R over MC1R, MC3R, and MC5R (Kis = 0.55, 16.78, 56.79, and >500 nM, respectively); decreases food intake and body weight in a rat model of diet-induced obesity at 0.075 or 0.299 µmol/kg
Catalog No. 50-294-3038 Supplier Cayman Chemical Supplier No. 4146910 MG
An MC4R agonist; induces cAMP release in HEK293 cells expressing human MC4R (EC50 = 0.28 nM); is selective for MC4R over MC1R, MC3R, and MC5R (Kis = 0.55, 16.78, 56.79, and >500 nM, respectively); decreases food intake and body weight in a rat model of diet-induced obesity at 0.075 or 0.299 µmol/kg