Learn More
Cayman Chemical Ac-DEVD-AFC 10mg
SDP
Supplier:  Cayman Chemical 1445910 MG
Encompass_Preferred
A fluorogenic substrate for activated caspase-3 (Km = 9.7 µM), as well as related caspases; caspase activity can be quantified by fluorescent detection of free AFC, which is excited at 400 nm and emits at 505 nm
Catalog No. 50-297-0005