Learn More
Medchemexpress LLC HY-P1005 1mg Medchemexpress, Ac-DEVD-AFC CAS:201608-14-2 Purity:>98%
SDP
Catalog No. 501934125
Encompass_Preferred
Medchemexpress, HY-P1005 1mg Ac-DEVD-AFC CAS:201608-14-2 Ac-DEVD-AFC is a fluorogenic substrate (λ ex =400 nm, λ em =530 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4125 Supplier Medchemexpress LLC Supplier No. HYP10051MG
Medchemexpress, HY-P1005 1mg Ac-DEVD-AFC CAS:201608-14-2 Ac-DEVD-AFC is a fluorogenic substrate (λ ex =400 nm, λ em =530 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.