Learn More
Medchemexpress LLC Ac-DEVD-AFC | 201608-14-2 | 98.9% | 729.61 | 10 MG
SDP
Catalog No. 5000424043
Encompass_Preferred
Ac-DEVD-AFC is a fluorogenic substrate designed for detecting caspase-3 activity, exhibiting specific excitation and emission wavelengths (λex=400 nm, λem=530 nm). It is characterized by the sequence N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoroMethylcoumarin and is suitable for various laboratory research applications. Significant increases in caspase-3 activity can be observed after incubation.
  • Fluorogenic substrate with λex=400 nm and λem=530 nm.
  • Specifically detects caspase-3 activity.
  • Activity can be observed within 4 hours of incubation.
  • Cleavage of the substrate is inhibited by shRNA-BimL transfection.
  • Stable under recommended storage conditions for extended periods.
  • Available in powder form and as a solution for diverse experimental needs.
Catalog No. 50-004-24043 Supplier Medchemexpress LLC Supplier No. HYP100510MG
Ac-DEVD-AFC is a fluorogenic substrate designed for detecting caspase-3 activity, exhibiting specific excitation and emission wavelengths (λex=400 nm, λem=530 nm). It is characterized by the sequence N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoroMethylcoumarin and is suitable for various laboratory research applications. Significant increases in caspase-3 activity can be observed after incubation.
  • Fluorogenic substrate with λex=400 nm and λem=530 nm.
  • Specifically detects caspase-3 activity.
  • Activity can be observed within 4 hours of incubation.
  • Cleavage of the substrate is inhibited by shRNA-BimL transfection.
  • Stable under recommended storage conditions for extended periods.
  • Available in powder form and as a solution for diverse experimental needs.