Learn More
Cayman Chemical Ac-DEVD-AMC, Purity: ≥95%, CAS: 169332-61-0, 10 mg
SDP
Catalog No. 502972307
Encompass_Preferred
A fluorogenic substrate that can be cleaved specifically by caspase-3
Catalog No. 50-297-2307 Supplier Cayman Chemical Supplier No. 1498610 MG
A fluorogenic substrate that can be cleaved specifically by caspase-3