Learn More
Cayman Chemical Ac-DEVD-AMC, 169332-61-0, 10 mg
SDP
Supplier:  Cayman Chemical 1498610
Encompass_Preferred
Molecular Formula C30H37N5O13, Purity ≥95%, Formula Weight 675.6, Synonyms: N-acetyl-L-α-aspartyl-L-α-glutamyl-L-valyl-N-(4-methyl-2-oxo-2H-1-benzopyran-7-yl)-L-α-asparagine, N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-Methylcoumarin, Caspase-3 Substrate (Fluorogenic); A fluorogenic substrate that can be cleaved specifically by caspase-3.
Catalog No. NC1399305