Learn More
Medchemexpress LLC Ac-DEVD-AMC | 169332-61-0 | 99.18% | 675.64 | 50 MG
SDP
Catalog No. 5000423672
Encompass_Preferred
Ac-DEVD-AMC is a fluorescent substrate of caspase-3/caspase-7. When treated with cell lysate, Ac-DEVD-AMC releases amino-4-methylcoumarin (AMC) for fluorescence detection. It has an excitation wavelength of 380 nm and an emission wavelength of 460 nm.
  • Fluorescent substrate for caspase-3/caspase-7.
  • Releases amino-4-methylcoumarin (AMC) for fluorescence detection.
  • Excitation wavelength: 380 nm.
  • Emission wavelength: 460 nm.
  • For research use only.
Catalog No. 50-004-23672 Supplier Medchemexpress LLC Supplier No. HYP100350MG
Ac-DEVD-AMC is a fluorescent substrate of caspase-3/caspase-7. When treated with cell lysate, Ac-DEVD-AMC releases amino-4-methylcoumarin (AMC) for fluorescence detection. It has an excitation wavelength of 380 nm and an emission wavelength of 460 nm.
  • Fluorescent substrate for caspase-3/caspase-7.
  • Releases amino-4-methylcoumarin (AMC) for fluorescence detection.
  • Excitation wavelength: 380 nm.
  • Emission wavelength: 460 nm.
  • For research use only.