Learn More
Medchemexpress LLC Ac-DEVD-AMC | 169332-61-0 | 99.2% | 675.64 | 100 MG
SDP
Catalog No. 5000424514
Encompass_Preferred
Ac-DEVD-AMC is a fluorescent substrate designed for caspase-3 and caspase-7. When incubated with cell lysate, it releases amino-4-methylcoumarin (AMC), enabling fluorescence detection at specific wavelengths.
  • Fluorescent substrate for caspase-3/caspase-7
  • Releases amino-4-methylcoumarin (AMC) for fluorescence detection
  • Excitation wavelength of 380 nm
  • Emission wavelength of 460 nm
  • For research use only
Catalog No. 50-004-24514 Supplier Medchemexpress LLC Supplier No. HYP1003100MG
Ac-DEVD-AMC is a fluorescent substrate designed for caspase-3 and caspase-7. When incubated with cell lysate, it releases amino-4-methylcoumarin (AMC), enabling fluorescence detection at specific wavelengths.
  • Fluorescent substrate for caspase-3/caspase-7
  • Releases amino-4-methylcoumarin (AMC) for fluorescence detection
  • Excitation wavelength of 380 nm
  • Emission wavelength of 460 nm
  • For research use only