Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-DEVD-AMC | 169332-61-0 | MFCD00671411 | 99.2% | 675.64 | 5 MG
SDP

Catalog No. 5000424554
Encompass_Preferred

Ac-DEVD-AMC is a fluorescent substrate of caspase-3/caspase-7. When treated with cell lysate, it releases amino-4-methylcoumarin (AMC) for fluorescence detection, with an excitation wavelength of 380 nm and an emission wavelength of 460 nm.

  • Purity 99.18%
  • Molecular weight 675.64
  • Appearance as a solid
  • Color white to off-white
  • Emission wavelength 480 nm
  • Excitation wavelength 390 nm
  • Recommended powder storage: -80°C for 2 years, -20°C for 1 year
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • In vitro solubility in DMSO: 10 mg/mL (14.80 mM)

Catalog No. 50-004-24554 Supplier Medchemexpress LLC Supplier No. HYP10035MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-DEVD-AMC is a fluorescent substrate of caspase-3/caspase-7. When treated with cell lysate, it releases amino-4-methylcoumarin (AMC) for fluorescence detection, with an excitation wavelength of 380 nm and an emission wavelength of 460 nm.

  • Purity 99.18%
  • Molecular weight 675.64
  • Appearance as a solid
  • Color white to off-white
  • Emission wavelength 480 nm
  • Excitation wavelength 390 nm
  • Recommended powder storage: -80°C for 2 years, -20°C for 1 year
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • In vitro solubility in DMSO: 10 mg/mL (14.80 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.