Learn More
Cayman Chemical Ac-DEVD-AMC, Purity: ≥95%, CAS: 169332-61-0, 1 mg
SDP
Catalog No. 502971268
Encompass_Preferred
A fluorogenic substrate that can be cleaved specifically by caspase-3
Catalog No. 50-297-1268 Supplier Cayman Chemical Supplier No. 149861 MG
A fluorogenic substrate that can be cleaved specifically by caspase-3