Learn More
TARGETMOL CHEMICALS INC Ac-DEVD-AMC 5MG
SDP
Catalog No. 502878316
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ac-DEVD-AMC (AC-ASP-MET-GLN-ASP-7-AMINO-4-METHYLCOUMARIN) is the substrate of Caspase-3 . Purity 98.2%
Catalog No. 50-287-8316 Supplier TARGETMOL CHEMICALS INC Supplier No. T189095MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ac-DEVD-AMC (AC-ASP-MET-GLN-ASP-7-AMINO-4-METHYLCOUMARIN) is the substrate of Caspase-3 . Purity 98.2%