Learn More
Medchemexpress LLC Ac-DEVD-CHO | 169332-60-9 | 99.76% | 502.47 | 10 MG
SDP
Catalog No. 5000424512
Encompass_Preferred
Ac-DEVD-CHO is a Caspase-3 inhibitor with a Ki value of 230 pM. It is for research use only and not sold to patients.
  • Inhibits Caspase-3 with a Ki value of 230 pM.
  • Also inhibits Caspase-8 (Ki: 0.92 nM), Caspase-7 (Ki: 1.6 nM), Caspase-10 (Ki: 12 nM), Caspase-1 (Ki: 18 nM), Caspase-6 (Ki: 31 nM), Caspase-9 (Ki: 60 nM), Caspase-4 (Ki: 132 nM), Caspase-5 (Ki: 205 nM), and Caspase-2 (Ki: 1710 nM).
  • Significantly prevents SLNT-induced apoptosis.
  • Partially blocks SIN-induced apoptosis and reduces the number of apoptotic nuclei.
Catalog No. 50-004-24512 Supplier Medchemexpress LLC Supplier No. HYP100110MG
Ac-DEVD-CHO is a Caspase-3 inhibitor with a Ki value of 230 pM. It is for research use only and not sold to patients.
  • Inhibits Caspase-3 with a Ki value of 230 pM.
  • Also inhibits Caspase-8 (Ki: 0.92 nM), Caspase-7 (Ki: 1.6 nM), Caspase-10 (Ki: 12 nM), Caspase-1 (Ki: 18 nM), Caspase-6 (Ki: 31 nM), Caspase-9 (Ki: 60 nM), Caspase-4 (Ki: 132 nM), Caspase-5 (Ki: 205 nM), and Caspase-2 (Ki: 1710 nM).
  • Significantly prevents SLNT-induced apoptosis.
  • Partially blocks SIN-induced apoptosis and reduces the number of apoptotic nuclei.