Learn More
Medchemexpress LLC HY-P1001 1mg Medchemexpress, Ac-DEVD-CHO CAS:169332-60-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP10011MG
Encompass_Preferred
Medchemexpress, HY-P1001 1mg Ac-DEVD-CHO CAS:169332-60-9 Ac-DEVD-CHO is a specific Caspase-3 inhibitor with a K i value of 230 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4122