Learn More
Selleck Chemical LLC Ac-DEVD-CHO 2mg 169332-60-9 Caspase-3 Inhibitor I, N-Ac-Asp-Glu-Val-Asp-CHO
SDP
Catalog No. 501365390
Encompass_Preferred
Ac-DEVD-CHO (Caspase-3 Inhibitor I, N-Ac-Asp-Glu-Val-Asp-CHO) is a potent aldehyde inhibitor of Group II caspases with Ki values of 0.2 nM and 0.3 nM for for caspase-3 and caspase-7, respectively. Weak inhibition for caspase-2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5390 Supplier Selleck Chemical LLC Supplier No. S79012MG
Ac-DEVD-CHO (Caspase-3 Inhibitor I, N-Ac-Asp-Glu-Val-Asp-CHO) is a potent aldehyde inhibitor of Group II caspases with Ki values of 0.2 nM and 0.3 nM for for caspase-3 and caspase-7, respectively. Weak inhibition for caspase-2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.