Learn More
Biotium Ac-DEVD-CHO, Caspase-3 Inhibitor
Catalog No. 501964125
Encompass_Preferred
Ac-DEVD-CHO is a synthetic tetrapeptide competitive inhibitor for Caspase-3/7, and contains the amino acid sequence of the PARP cleavage site. Ac-DEVD-CHO can be used to inhibit Caspase-3/7 activity in apoptotic cells, and to study events downstream of Caspase-3/7 activation.Properties: Soluble in DMSO; Store at 4°C; C20H30N4O11; MW: 502.5.
Catalog No. 50-196-4125 Supplier Biotium Supplier No. 10404
Ac-DEVD-CHO is a synthetic tetrapeptide competitive inhibitor for Caspase-3/7, and contains the amino acid sequence of the PARP cleavage site. Ac-DEVD-CHO can be used to inhibit Caspase-3/7 activity in apoptotic cells, and to study events downstream of Caspase-3/7 activation.Properties: Soluble in DMSO; Store at 4°C; C20H30N4O11; MW: 502.5.