Learn More
CPC Scientific Ac-Asp-Glu-Val-Asp-CMK 1MG
SDP
Catalog No. 502109008
Encompass
ONE-LETTER SEQUENCE: Ac-DEVD-CMKMOLECULAR FORMULA: C21H31N4O11Cl1MOLECULAR WEIGHT:550.95STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [285570-60-7], SYNONYMS: Ac-Asp-Glu-Val-Asp-chloromethylketoneRESEARCH AREA: Apoptosis
Catalog No. 50-210-9008 Supplier CPC Scientific Supplier No. CASP051A
ONE-LETTER SEQUENCE: Ac-DEVD-CMKMOLECULAR FORMULA: C21H31N4O11Cl1MOLECULAR WEIGHT:550.95STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [285570-60-7], SYNONYMS: Ac-Asp-Glu-Val-Asp-chloromethylketoneRESEARCH AREA: Apoptosis