Learn More
CPC Scientific Ac-Asp-Glu-Val-Asp-CMK 5MG
SDP
Catalog No. 502109009
Encompass
ONE-LETTER SEQUENCE: Ac-DEVD-CMKMOLECULAR FORMULA: C21H31N4O11Cl1MOLECULAR WEIGHT:550.95STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [285570-60-7], SYNONYMS: Ac-Asp-Glu-Val-Asp-chloromethylketoneRESEARCH AREA: Apoptosis
Catalog No. 50-210-9009 Supplier CPC Scientific Supplier No. CASP051B
Add to Cart
Add to Cart
ONE-LETTER SEQUENCE: Ac-DEVD-CMKMOLECULAR FORMULA: C21H31N4O11Cl1MOLECULAR WEIGHT:550.95STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [285570-60-7], SYNONYMS: Ac-Asp-Glu-Val-Asp-chloromethylketoneRESEARCH AREA: Apoptosis