Learn More
Medchemexpress LLC Ac-devd-cmk tfa | 98.6% | 664.97 | 5 MG
SDP
Catalog No. 5000426511
Encompass_Preferred
Ac-DEVD-CMK TFA is a selective and irreversible caspase-3 inhibitor. It significantly inhibits apoptosis induced by high levels of glucose or 3,20-dibenzoate, and can be used in various experimental approaches to inhibit apoptosis.
  • Selective and irreversible caspase-3 inhibitor.
  • Significantly inhibits apoptosis induced by high levels of glucose or 3,20-dibenzoate.
  • Suitable for various experimental approaches to inhibit apoptosis.
  • Inhibits IDB-induced apoptosis.
  • Attenuates acetaminophen-induced liver injury.
  • Mitigates liver damage by inhibiting GSK‐3β or caspase‐3 activity.
  • Effective in inhibiting citrate-induced p21 cleavage and G2/M accumulation in human pharyngeal squamous carcinoma cell lines.
Catalog No. 50-004-26511 Supplier Medchemexpress LLC Supplier No. HYP0034A5MG
Ac-DEVD-CMK TFA is a selective and irreversible caspase-3 inhibitor. It significantly inhibits apoptosis induced by high levels of glucose or 3,20-dibenzoate, and can be used in various experimental approaches to inhibit apoptosis.
  • Selective and irreversible caspase-3 inhibitor.
  • Significantly inhibits apoptosis induced by high levels of glucose or 3,20-dibenzoate.
  • Suitable for various experimental approaches to inhibit apoptosis.
  • Inhibits IDB-induced apoptosis.
  • Attenuates acetaminophen-induced liver injury.
  • Mitigates liver damage by inhibiting GSK‐3β or caspase‐3 activity.
  • Effective in inhibiting citrate-induced p21 cleavage and G2/M accumulation in human pharyngeal squamous carcinoma cell lines.