Learn More
CPC Scientific Ac-Asp-Glu-Val-Asp-pNA 1MG
SDP
Supplier:  CPC Scientific CASP052A
Encompass
Cleavage of this chromogenic peptide substrate for caspase-3 is detected at a wavelength of 405 nm. ONE-LETTER SEQUENCE: Ac-DEVD-pNAMOLECULAR FORMULA: C26H34N6O13MOLECULAR WEIGHT:638.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [189950-66-1], RESEARCH AREA: ApoptosisREFERENCES: D.W. Nicholson, Nature, 376 (1995)
Catalog No. 50-210-9010