Learn More
Enzo Life Sciences Ac-DEVD-pNA (5mg)
SDP
Supplier:  Enzo Life Sciences ALX260033M005
Encompass_Preferred
Colorimetric substrate for caspase-3 (CPP32) (Km=9.7µM) and related cysteine proteases. Sequence is based on PARP (poly(ADPribose) polymerase) cleavage site Asp216 for caspase-3. Cleavage is monitored colorimetrically at 405nm. Alternative name: Caspase-3 substrate (chromogenic). Formula: C26H34N6O13. MW: 638.6. Purity: ≥97% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO or buffer, pH 7. Long Term Storage: -20°C.
Catalog No. 50-200-8245