Learn More
Apexbio Technology LLC Ac-Endothelin-1 (16-21), human 1mg
SDP
Catalog No. 5000092802
Encompass_Preferred
Ac-Endothelin-1 16-21 human is a selective agonist targeting the ETB subtype of G protein-coupled endothelin receptors It is designed to activate ETB receptors thereby modulating phospholipase C-mediated signaling pathways and vascular function Ac-Endothelin-1 16-21 human exerts its biological activity primarily through selective ETB receptor activation leading to vasoconstrictive effects Based on these pharmacological properties Ac-Endothelin-1 16-21 human holds research potential in studies analyzing vascular function as well as investigations of cardiovascular and renal pathophysiology
Catalog No. 50-000-92802 Supplier Apexbio Technology LLC Supplier No. A10161
Ac-Endothelin-1 16-21 human is a selective agonist targeting the ETB subtype of G protein-coupled endothelin receptors It is designed to activate ETB receptors thereby modulating phospholipase C-mediated signaling pathways and vascular function Ac-Endothelin-1 16-21 human exerts its biological activity primarily through selective ETB receptor activation leading to vasoconstrictive effects Based on these pharmacological properties Ac-Endothelin-1 16-21 human holds research potential in studies analyzing vascular function as well as investigations of cardiovascular and renal pathophysiology