Learn More
Medchemexpress LLC Ac-Exatecan | 2922852-48-8 | 99.0% | C26H24FN3O5 | 1 ML
SDP
Catalog No. 5000433015
Encompass_Preferred
Ac-Exatecan is an acetylation-modified Exatecan, which functions as a common toxin component in Antibody-Drug Conjugate (ADC) preparation. It acts as an inhibitor of DNA topoisomerase I, with an IC50 of 2.2 μM, and demonstrates antitumor activity. This compound can be utilized as an ADC payload and is effective against P-glycoprotein mediated multi-drug resistant cells.
  • Acetylation-modified Exatecan
  • Functions as a common toxin component in ADC preparation (ADC cytotoxin)
  • Inhibits DNA topoisomerase I (IC50 = 2.2 μM)
  • Exhibits antitumor activity
  • Can be utilized as a payload for ADC
  • Effective against P-glycoprotein mediated multi-drug resistant cells
Catalog No. 50-004-33015 Supplier Medchemexpress LLC Supplier No. HY7949010MM/1ML
Ac-Exatecan is an acetylation-modified Exatecan, which functions as a common toxin component in Antibody-Drug Conjugate (ADC) preparation. It acts as an inhibitor of DNA topoisomerase I, with an IC50 of 2.2 μM, and demonstrates antitumor activity. This compound can be utilized as an ADC payload and is effective against P-glycoprotein mediated multi-drug resistant cells.
  • Acetylation-modified Exatecan
  • Functions as a common toxin component in ADC preparation (ADC cytotoxin)
  • Inhibits DNA topoisomerase I (IC50 = 2.2 μM)
  • Exhibits antitumor activity
  • Can be utilized as a payload for ADC
  • Effective against P-glycoprotein mediated multi-drug resistant cells