Learn More
Medchemexpress LLC Ac-Exatecan | 2922852-48-8 | 99.0% | C26H24FN3O5 | 50 MG
SDP
Catalog No. 5000433255
Encompass_Preferred
Ac-Exatecan is an acetylation-modified Exatecan, which acts as a common toxin component in Antibody-Drug Conjugate (ADC) preparation and is an inhibitor of DNA topoisomerase I (IC50 = 2.2 μM). This compound demonstrates antitumor activity and can serve as a payload for ADCs. Furthermore, Exatecan is effective against P-glycoprotein-mediated multi-drug resistant cells. This product is for research use only.
  • Acetylation-modified Exatecan
  • Common toxin component in antibody-drug conjugate (ADC) preparation
  • Inhibitor of DNA topoisomerase I (IC50 = 2.2 μM)
  • Demonstrates antitumor activity
  • Can serve as a payload for ADCs
  • Effective against P-glycoprotein-mediated multi-drug resistant cells
Catalog No. 50-004-33255 Supplier Medchemexpress LLC Supplier No. HY7949050MG
Ac-Exatecan is an acetylation-modified Exatecan, which acts as a common toxin component in Antibody-Drug Conjugate (ADC) preparation and is an inhibitor of DNA topoisomerase I (IC50 = 2.2 μM). This compound demonstrates antitumor activity and can serve as a payload for ADCs. Furthermore, Exatecan is effective against P-glycoprotein-mediated multi-drug resistant cells. This product is for research use only.
  • Acetylation-modified Exatecan
  • Common toxin component in antibody-drug conjugate (ADC) preparation
  • Inhibitor of DNA topoisomerase I (IC50 = 2.2 μM)
  • Demonstrates antitumor activity
  • Can serve as a payload for ADCs
  • Effective against P-glycoprotein-mediated multi-drug resistant cells