Learn More
Medchemexpress LLC Ac-exatecan-d5 | 2922852-48-8 | 99.9% | 482.52 | C26H19D5FN3O5 | 10 MG
SDP
Catalog No. 5000299106
Encompass_Preferred
Ac-Exatecan-d5 is a deuterium-labeled analogue of Ac-Exatecan provided for research use as a tracer and internal standard in quantitative analyses. It is supplied as a high-purity solid and is intended for laboratory analytical applications requiring isotopically labeled standards.
  • Deuterium-labeled tracer for quantitative analysis.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity (approx. 99.9%).
  • Solid, light yellow to yellow appearance.
  • Recommended storage: sealed and away from moisture; in solvent, -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-002-99106 Supplier Medchemexpress LLC Supplier No. HY79490S10MG
Add to Cart
Add to Cart
Ac-Exatecan-d5 is a deuterium-labeled analogue of Ac-Exatecan provided for research use as a tracer and internal standard in quantitative analyses. It is supplied as a high-purity solid and is intended for laboratory analytical applications requiring isotopically labeled standards.
  • Deuterium-labeled tracer for quantitative analysis.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity (approx. 99.9%).
  • Solid, light yellow to yellow appearance.
  • Recommended storage: sealed and away from moisture; in solvent, -80°C for 6 months or -20°C for 1 month.