Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-Exatecan-d5 | 2922852-48-8 | 99.9% | C26H19D5FN3O5 | 25 MG
SDP

Catalog No. 5000357532
Encompass_Preferred

Ac-Exatecan-d5 is the deuterium-labeled version of Ac-Exatecan. This compound is intended for research use only and is not sold to patients.

  • Can be used as a tracer and as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Deuterium substitution in drug molecules is largely used as a tracer for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Molecular weight: 482.52.
  • Appearance: Solid, light yellow to yellow.
  • Ships at room temperature in the continental US (may vary elsewhere). Store at 4°C, sealed, away from moisture. In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture).

Catalog No. 50-003-57532 Supplier Medchemexpress LLC Supplier No. 25MGHY79490S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-Exatecan-d5 is the deuterium-labeled version of Ac-Exatecan. This compound is intended for research use only and is not sold to patients.

  • Can be used as a tracer and as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Deuterium substitution in drug molecules is largely used as a tracer for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Molecular weight: 482.52.
  • Appearance: Solid, light yellow to yellow.
  • Ships at room temperature in the continental US (may vary elsewhere). Store at 4°C, sealed, away from moisture. In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.