Learn More
Medchemexpress LLC Ac-Exatecan-d5 | 2922852-48-8 | 99.9% | C26H19D5FN3O5 | 5 MG
SDP
Catalog No. 5000433268
Encompass_Preferred
Ac-Exatecan-d5 is a deuterium-labeled version of Ac-Exatecan, intended for research use only. It can be used as a tracer or an internal standard for quantitative analysis, leveraging stable heavy isotopes like deuterium for incorporation into drug molecules during development. Deuteration may influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can serve as an internal standard for quantitative analysis
  • Suitable for techniques such as NMR, GC-MS, or LC-MS
Catalog No. 50-004-33268 Supplier Medchemexpress LLC Supplier No. HY79490S5MG
Add to Cart
Add to Cart
Ac-Exatecan-d5 is a deuterium-labeled version of Ac-Exatecan, intended for research use only. It can be used as a tracer or an internal standard for quantitative analysis, leveraging stable heavy isotopes like deuterium for incorporation into drug molecules during development. Deuteration may influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can serve as an internal standard for quantitative analysis
  • Suitable for techniques such as NMR, GC-MS, or LC-MS