Learn More
Selleck Chemical LLC Ac-FLTD-CMK S9817-25MG
SDP
Catalog No. 5000024688
Encompass_Preferred
Ac-FLTD-CMK is a specific inhibitor for inflammatory caspases Ac-FLTD-CMK inhibits gasdermin D (GSDMD) cleavage by caspases-1 -4 -5 and -11 in vitro Ac-FLTD-CMK is effective against caspases-1 caspases-4 and caspases-5 with IC50 of 46 7 nM 1 49 M and 329 nM respectively
Catalog No. 50-000-24688 Supplier Selleck Chemical LLC Supplier No. S981725MG
Ac-FLTD-CMK is a specific inhibitor for inflammatory caspases Ac-FLTD-CMK inhibits gasdermin D (GSDMD) cleavage by caspases-1 -4 -5 and -11 in vitro Ac-FLTD-CMK is effective against caspases-1 caspases-4 and caspases-5 with IC50 of 46 7 nM 1 49 M and 329 nM respectively