Learn More
TARGETMOL CHEMICALS INC AC-FLTD-CMK 5MG
SDP
Catalog No. 502580167
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Ac-FLTD-CMK is an inhibitor derived from abscisicin D (GSDMDD) with specific inhibitory effects on inflammatory caspases. ac-FLTD-CMK showed inhibitory effects on caspases-1 caspases-4 and caspases-11 with IC50s of 46.7 nM 1.49 uM and 329 nM respectively. Ac-FLTD-CMK showed potency against caspases-1 caspases-4 and caspases-11 but not against apoptosis-associated caspase-3. purity: 95%
Catalog No. 50-258-0167 Supplier TARGETMOL CHEMICALS INC Supplier No. T368195MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Ac-FLTD-CMK is an inhibitor derived from abscisicin D (GSDMDD) with specific inhibitory effects on inflammatory caspases. ac-FLTD-CMK showed inhibitory effects on caspases-1 caspases-4 and caspases-11 with IC50s of 46.7 nM 1.49 uM and 329 nM respectively. Ac-FLTD-CMK showed potency against caspases-1 caspases-4 and caspases-11 but not against apoptosis-associated caspase-3. purity: 95%