Learn More
Bachem Ac-Gly-Ala-Lys(Ac)-AMC
SDP
Catalog No. 50260362
Encompass_Preferred
Ac-Gly-Ala-Lys(Ac)-AMC, 577969-56-3, C25H33N5O7, Mr 515.57, 5mg. Ac-GAK(Ac)-AMC, fluorogenic substrate for measuring histone deacetylase class I (HDAC 1, 2, 3, and 8) and class II (HDAC 6 and 10) activity in a protease-coupled assay. HDAC-catalyzed deacetylation of Lys yields Ac-GAK-AM.
Catalog No. 50-260-362 Supplier Bachem Supplier No. 4060671.0005
Ac-Gly-Ala-Lys(Ac)-AMC, 577969-56-3, C25H33N5O7, Mr 515.57, 5mg. Ac-GAK(Ac)-AMC, fluorogenic substrate for measuring histone deacetylase class I (HDAC 1, 2, 3, and 8) and class II (HDAC 6 and 10) activity in a protease-coupled assay. HDAC-catalyzed deacetylation of Lys yields Ac-GAK-AM.