Learn More
CPC Scientific Ac-Gly-Ala-Lys(Ac)-AMC 25MG
SDP
Supplier:  CPC Scientific HIST007B
Encompass
This peptide is the fluorogenic substrate for measuring histone deacetylase class I (HDAC 1, 2, 3, and 8) and class II (HDAC 6 and 10) activity in a protease-coupled assay. ONE-LETTER SEQUENCE: Ac-GAK(Ac)-AmcMOLECULAR FORMULA: C25H33N5O7MOLECULAR WEIGHT:515.57STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [577969-56-3], RESEARCH AREA: MiscellaneousREFERENCES: B.E.Schultz et al., Biochemistry, 43, 11083 (2004)
Catalog No. 50-210-9435