Learn More
Cayman Chemical Ac-Gly-Lys-OMe (acetate), Purity: ≥95%, CAS: 14752-92-2, 10 mg
SDP
Catalog No. 502992190
Encompass_Preferred
A synthetic peptide urokinase substrate; has been used to characterize the effects of small molecule inhibitors and metal ions on urokinase activity in cell-free assays
Catalog No. 50-299-2190 Supplier Cayman Chemical Supplier No. 3056810 MG
A synthetic peptide urokinase substrate; has been used to characterize the effects of small molecule inhibitors and metal ions on urokinase activity in cell-free assays