Learn More
Enzo Life Sciences Ac-Gly-Pro-Leu-Asp-AMC (5mg)
SDP
Catalog No. 502010334
Encompass_Preferred
Highly specific substrate for the caspase-like peptidic activity of the 20S proteasome. Alternative name: Ac-GPLD-AMC. Purity: ≥98% (HPLC). Solubility: Soluble in 20% aqueous acetic acid (1mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0334 Supplier Enzo Life Sciences Supplier No. BMLAW95600005
Highly specific substrate for the caspase-like peptidic activity of the 20S proteasome. Alternative name: Ac-GPLD-AMC. Purity: ≥98% (HPLC). Solubility: Soluble in 20% aqueous acetic acid (1mg/ml). Long Term Storage: -20°C.