Learn More
Medchemexpress LLC Ac-GpYLPQTV-NH2 acetate | 99.38% | 897.91 | 10 MG
SDP
Catalog No. 5000422587
Encompass_Preferred
Ac-GpYLPQTV-NH2 acetate is a STAT3 inhibitor with an IC50 value of 0.33 μM and has antitumor activity.
  • STAT3 inhibitor
  • Antitumor activity
Catalog No. 50-004-22587 Supplier Medchemexpress LLC Supplier No. HYP10201A10MG
Ac-GpYLPQTV-NH2 acetate is a STAT3 inhibitor with an IC50 value of 0.33 μM and has antitumor activity.
  • STAT3 inhibitor
  • Antitumor activity