Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-GpYLPQTV-NH2 acetate | 99.4% | 897.91 (free base) | 1 MG
SDP

Catalog No. 5000423713
Encompass_Preferred

Ac-GpYLPQTV-NH2 acetate is a STAT3 inhibitor with an IC50 value of 0.33 μM. It exhibits antitumor activity and is intended for research use only.

  • STAT3 inhibitor (IC50: 0.33 μM)
  • Exhibits antitumor activity
  • White to off-white solid appearance
  • Soluble in DMSO (100 mg/mL)
  • Storage as powder: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture)
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-004-23713 Supplier Medchemexpress LLC Supplier No. HYP10201A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-GpYLPQTV-NH2 acetate is a STAT3 inhibitor with an IC50 value of 0.33 μM. It exhibits antitumor activity and is intended for research use only.

  • STAT3 inhibitor (IC50: 0.33 μM)
  • Exhibits antitumor activity
  • White to off-white solid appearance
  • Soluble in DMSO (100 mg/mL)
  • Storage as powder: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture)
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.