Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-green (VDP-green) | 2937705-58-1 | 98.0% | 10 MG
SDP

Catalog No. 5000298136
Encompass_Preferred

AC-green (VDP-green) is a fluorescent β-allyl carbamate probe for selective detection and imaging of vicinal dithiol proteins in living systems. It offers high sensitivity and low toxicity for cellular imaging and can detect reduced bovine serum albumin (rBSA).

  • Selective imaging of vicinal dithiol proteins in cells.
  • Low toxicity and high sensitivity for biological imaging.
  • Excitation/emission approximately 400 nm/475 nm.
  • Chemical formula C24H25N3O5, molecular weight 435.47 g/mol.
  • Purity approximately 98.0% and soluble in DMSO at 100 mg/mL.
  • Storage: store at -20°C protected from light; in solvent store at -80°C for long-term.

Catalog No. 50-002-98136 Supplier Medchemexpress LLC Supplier No. HYD125810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AC-green (VDP-green) is a fluorescent β-allyl carbamate probe for selective detection and imaging of vicinal dithiol proteins in living systems. It offers high sensitivity and low toxicity for cellular imaging and can detect reduced bovine serum albumin (rBSA).

  • Selective imaging of vicinal dithiol proteins in cells.
  • Low toxicity and high sensitivity for biological imaging.
  • Excitation/emission approximately 400 nm/475 nm.
  • Chemical formula C24H25N3O5, molecular weight 435.47 g/mol.
  • Purity approximately 98.0% and soluble in DMSO at 100 mg/mL.
  • Storage: store at -20°C protected from light; in solvent store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.