Learn More
Climet Instruments Climet CI-3100 OPT Particle Counter, 2-Channel, 1 CFM Flow Rate, VHP Compatible
SDP
Catalog No. NC3589864
Encompass
The CI-3100 OPT is a biopharmaceutical-grade particle counter designed for precise continuous air quality monitoring in cleanroom environments. It features dual channels for 0.5 µm and 5.0 µm particles, a flow rate of 1 CFM, and 4-20 mA output for easy system integration. Its durable stainless steel enclosure and internal components eliminate external air source needs, ensuring reliable performance and compliance with industry standards. Ideal for critical applications, it enhances operational efficiency in maintaining cleanroom integrity.
Catalog No. NC3589864 Supplier Climet Instruments Supplier No. CI310012144
Add to Cart
Add to Cart
Request a Quote
The CI-3100 OPT is a biopharmaceutical-grade particle counter designed for precise continuous air quality monitoring in cleanroom environments. It features dual channels for 0.5 µm and 5.0 µm particles, a flow rate of 1 CFM, and 4-20 mA output for easy system integration. Its durable stainless steel enclosure and internal components eliminate external air source needs, ensuring reliable performance and compliance with industry standards. Ideal for critical applications, it enhances operational efficiency in maintaining cleanroom integrity.