Learn More
Cayman Chemical Ac-IEPD-AFC 5mg
SDP
Catalog No. 502978441
Encompass_Preferred
A fluorogenic substrate for granzyme B; AFC is released upon enzymatic cleavage by granzyme B and its fluorescence can be used to quantify granzyme B activity; ex/em = 400/505 nm, respectively
Catalog No. 50-297-8441 Supplier Cayman Chemical Supplier No. 421795 MG
A fluorogenic substrate for granzyme B; AFC is released upon enzymatic cleavage by granzyme B and its fluorescence can be used to quantify granzyme B activity; ex/em = 400/505 nm, respectively