Learn More
Enzo Life Sciences Ac-IEPD-AMC (5mg)
SDP
Catalog No. 502011244
Encompass_Preferred
Fluorogenic substrate for caspase-8 and granzyme B. Ex.: 360 nm, Em.: 450 nm. The Km of recombinant human granzyme B for Ac-IEPD-AMC is 160 µM, with a Vmax about eight-fold lower than that of Ac-IEPD-pNA (BML-P133). Alternative name: Caspase 8 substrate (fluorogenic), Granzyme B substrate (fluorogenic). Purity: ≥95%. Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1244 Supplier Enzo Life Sciences Supplier No. BMLP1340005
Fluorogenic substrate for caspase-8 and granzyme B. Ex.: 360 nm, Em.: 450 nm. The Km of recombinant human granzyme B for Ac-IEPD-AMC is 160 µM, with a Vmax about eight-fold lower than that of Ac-IEPD-pNA (BML-P133). Alternative name: Caspase 8 substrate (fluorogenic), Granzyme B substrate (fluorogenic). Purity: ≥95%. Solubility: Soluble in DMSO. Long Term Storage: -20°C.