Learn More
Medchemexpress LLC Ac-IEPD-CHO | 352520-90-2 | 99.2% | 498.53 | 10 MG
SDP
Catalog No. 5000424422
Encompass_Preferred
Ac-IEPD-CHO is a reversible inhibitor of granzyme B, with a reported Ki of 80 nM. It also acts as a caspase-8 inhibitor.
  • Reversible granzyme B inhibitor.
  • Ki of 80 nM against granzyme B.
  • Caspase-8 inhibitor.
  • In vitro studies show Ac-IEPD-CHO (35-50 μg/mL, 1 h pretreatment followed by 16 h) diminishes Th17-polarized cells induced MO3.13 cell death.
Catalog No. 50-004-24422 Supplier Medchemexpress LLC Supplier No. HYP1014910MG
Ac-IEPD-CHO is a reversible inhibitor of granzyme B, with a reported Ki of 80 nM. It also acts as a caspase-8 inhibitor.
  • Reversible granzyme B inhibitor.
  • Ki of 80 nM against granzyme B.
  • Caspase-8 inhibitor.
  • In vitro studies show Ac-IEPD-CHO (35-50 μg/mL, 1 h pretreatment followed by 16 h) diminishes Th17-polarized cells induced MO3.13 cell death.