Learn More
Medchemexpress LLC Ac-IEPD-CHO | 352520-90-2 | 99.21% | 498.53 | 1 MG
SDP
Catalog No. 5000422512
Encompass_Preferred
Ac-IEPD-CHO is a reversible granzyme B inhibitor with a Ki of 80 nM. It also acts as a caspase-8 inhibitor.
  • Reversible granzyme B inhibitor with a Ki of 80 nM
  • Caspase-8 inhibitor
  • Diminishes Th17-polarized cells induced MO3.13 cell death
Catalog No. 50-004-22512 Supplier Medchemexpress LLC Supplier No. HYP101491MG
Ac-IEPD-CHO is a reversible granzyme B inhibitor with a Ki of 80 nM. It also acts as a caspase-8 inhibitor.
  • Reversible granzyme B inhibitor with a Ki of 80 nM
  • Caspase-8 inhibitor
  • Diminishes Th17-polarized cells induced MO3.13 cell death