Learn More
Abcam Ac-IEPD-CHO, granzyme B and caspase-8 inhibitor, 1MG
Catalog No. 502215312
Encompass_Preferred
MW 498.5 Da, Purity >95%. Ac-IEPD-CHO, granzyme B and caspase-8 inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5312 Supplier Abcam Supplier No. AB1420301MG
MW 498.5 Da, Purity >95%. Ac-IEPD-CHO, granzyme B and caspase-8 inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement