Learn More
Enzo Life Sciences Ac-IEPD-pNA (5mg)
SDP
Catalog No. 502011243
Encompass_Preferred
Colorimetric granzyme B substrate. Combinatorial and kinetic studies have found the IEPD sequence to be optimal for cleavage of a tetrapeptide substrate by recombinant granzyme B, with a Km for Ac-IEPD-pNA of 57 µM. Cleavage of the substrate by granzyme B releases pNA, which can be detected by its absorbance at 405 nm. Alternative name: Caspase 8 substrate (chromogenic), Granzyme B substrate (chromogenic). Purity: ≥95%. Solubility: Soluble to at least 100mM in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1243 Supplier Enzo Life Sciences Supplier No. BMLP1330005
Colorimetric granzyme B substrate. Combinatorial and kinetic studies have found the IEPD sequence to be optimal for cleavage of a tetrapeptide substrate by recombinant granzyme B, with a Km for Ac-IEPD-pNA of 57 µM. Cleavage of the substrate by granzyme B releases pNA, which can be detected by its absorbance at 405 nm. Alternative name: Caspase 8 substrate (chromogenic), Granzyme B substrate (chromogenic). Purity: ≥95%. Solubility: Soluble to at least 100mM in DMSO. Long Term Storage: -20°C.