Learn More
Enzo Life Sciences Ac-IETD-AFC (10 mg)
SDP
Catalog No. NC1463941
Encompass_Preferred
Caspase and granzyme substrate. Fluorogenic substrate for caspase-8/caspase-3 processing enzyme and granzyme B. Also a substrate for caspase-10. The IETD sequence is based on caspase-3 proenzyme cleavage site. Alternative Name: Caspase 8 substrate (fluorogenic), Granzyme B substrate (fluorogenic). Formula: C31H38F3N5O12. MW: 729.7. Purity: ≥98% (HPLC). Sequence Ac-Ile-Glu-Thr-Asp-AFC (AFC=7-Amino-4-trifluoromethylcoumarin). Appearance: White to off-white powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Handling: Keep cool and dry.
Catalog No. NC1463941 Supplier Enzo Life Sciences Supplier No. ALX260110M010
Caspase and granzyme substrate. Fluorogenic substrate for caspase-8/caspase-3 processing enzyme and granzyme B. Also a substrate for caspase-10. The IETD sequence is based on caspase-3 proenzyme cleavage site. Alternative Name: Caspase 8 substrate (fluorogenic), Granzyme B substrate (fluorogenic). Formula: C31H38F3N5O12. MW: 729.7. Purity: ≥98% (HPLC). Sequence Ac-Ile-Glu-Thr-Asp-AFC (AFC=7-Amino-4-trifluoromethylcoumarin). Appearance: White to off-white powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Handling: Keep cool and dry.