Learn More
Medchemexpress LLC Ac-IETD-AFC | 211990-57-7 | 99.28% | 729.65 | 5 MG
SDP
Catalog No. 5000422109
Encompass_Preferred
Ac-IETD-AFC is a fluorogenic substrate for caspase-8, caspase-3, caspase-10, and granzyme B. It is intended for research use only.
  • Fluorogenic substrate
  • Substrate for caspase-8, caspase-3, caspase-10
  • Substrate for granzyme B
  • Not intended for human use
Catalog No. 50-004-22109 Supplier Medchemexpress LLC Supplier No. HYP11695MG
Ac-IETD-AFC is a fluorogenic substrate for caspase-8, caspase-3, caspase-10, and granzyme B. It is intended for research use only.
  • Fluorogenic substrate
  • Substrate for caspase-8, caspase-3, caspase-10
  • Substrate for granzyme B
  • Not intended for human use