Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-IETD-AFC | 211990-57-7 | 99.3% | 729.65 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYP116910MG

Encompass_Preferred

Ac-IETD-AFC is a fluorogenic substrate for caspases-8, -3, and -10, as well as granzyme B. This product is intended for research use only and is not sold to patients.

  • Fluorogenic substrate for caspase-8, caspase-3, caspase-10, and granzyme B
  • Suitable for enzyme labeling and fluorescence studies
  • Used in apoptosis and caspase research
  • Excitation wavelength less than 380 nm
  • Emission wavelength 496-570 nm (green)

Catalog No. 50-004-23292


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.