Learn More
Medchemexpress LLC Ac-IETD-AFC | 211990-57-7 | 99.3% | 729.65 | 25 MG
SDP
Catalog No. 5000427015
Encompass_Preferred
Ac-IETD-AFC is a fluorogenic substrate that acts on caspase-8, caspase-3, caspase-10, and granzyme B.
  • Fluorogenic substrate
  • Acts on caspase-8
  • Acts on caspase-3
  • Acts on caspase-10
  • Acts on granzyme B
Catalog No. 50-004-27015 Supplier Medchemexpress LLC Supplier No. 25MGHYP1169
Add to Cart
Add to Cart
Ac-IETD-AFC is a fluorogenic substrate that acts on caspase-8, caspase-3, caspase-10, and granzyme B.
  • Fluorogenic substrate
  • Acts on caspase-8
  • Acts on caspase-3
  • Acts on caspase-10
  • Acts on granzyme B