Learn More
CPC Scientific Ac-Ile-Glu-Thr-Asp-AFC 5MG
SDP
Catalog No. 502109101
Encompass
Specific substrate for caspase-8.SEQUENCE: Ac-Ile-Glu-Thr-Asp-AFCONE-LETTER SEQUENCE: Ac-IETD-AFCMOLECULAR FORMULA: C31H38F3N5O12MOLECULAR WEIGHT: 729.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [211990-57-7]SYNONYMS: N-Acetyl-Ile-Glu-Thr-Asp-7-amino-4-TrifluoromethylcoumarinRESEARCH AREA: ApoptosisREFERENCES:Reszka, P., Schulz, R., Methling, K., et al. ChemMedChem 5(1), 103-117 (2010).SKU(s): CASP-095A, CASP-095B
Catalog No. 50-210-9101 Supplier CPC Scientific Supplier No. CASP095B
Specific substrate for caspase-8.SEQUENCE: Ac-Ile-Glu-Thr-Asp-AFCONE-LETTER SEQUENCE: Ac-IETD-AFCMOLECULAR FORMULA: C31H38F3N5O12MOLECULAR WEIGHT: 729.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [211990-57-7]SYNONYMS: N-Acetyl-Ile-Glu-Thr-Asp-7-amino-4-TrifluoromethylcoumarinRESEARCH AREA: ApoptosisREFERENCES:Reszka, P., Schulz, R., Methling, K., et al. ChemMedChem 5(1), 103-117 (2010).SKU(s): CASP-095A, CASP-095B