Learn More
Enzo Life Sciences Ac-IETD-AMC (1 mg)
SDP
Catalog No. NC1448185
Encompass_Preferred
Caspase and granzyme substrate. Fluorogenic substrate for caspase-6 and -8 and granzyme B. Also a substrate for caspase-10. Alternative Name: Granzyme B substrate (fluorogenic), Caspase-8 substrate (fluorogenic). Formula: C31H41N5O12. MW: 675.7. Purity: ≥97% (HPLC). Sequence Ac-Ile-Glu-Thr-Asp-AMC (AMC=7-Amino-4-methylcoumarin). Appearance: White to off-white powder. Solubility: Soluble in DMSO. Dilute with buffer, pH 7.5. Long Term Storage: -20°C. Handling: Protect from light. Keep cool and dry.
Catalog No. NC1448185 Supplier Enzo Life Sciences Supplier No. ALX260042M001
Caspase and granzyme substrate. Fluorogenic substrate for caspase-6 and -8 and granzyme B. Also a substrate for caspase-10. Alternative Name: Granzyme B substrate (fluorogenic), Caspase-8 substrate (fluorogenic). Formula: C31H41N5O12. MW: 675.7. Purity: ≥97% (HPLC). Sequence Ac-Ile-Glu-Thr-Asp-AMC (AMC=7-Amino-4-methylcoumarin). Appearance: White to off-white powder. Solubility: Soluble in DMSO. Dilute with buffer, pH 7.5. Long Term Storage: -20°C. Handling: Protect from light. Keep cool and dry.